Invitrogen
Manufacturer Code:PA528035
Catalog # PIPA528035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 258 of Human HAAO |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-HAO; 3-HAO HAO 3-hydroxyanthranilate oxygenase 3-hydroxyanthranilic acid dioxygenase HAD; 3-hydroxyanthranilate 3,4-dioxygenase; 3-hydroxyanthranilate oxygenase; 3-hydroxyanthranilic acid dioxygenase; HAAO; HAD
Gene Aliases: 0610007K21Rik; 0610012J07Rik; 3-HAO; 3-HAOxase; 3HAO; HAAO; HAO
UniProt ID: (Human) P46952, (Mouse) Q78JT3
Entrez Gene ID: (Human) 23498, (Mouse) 107766
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.