Novus Biologicals
Manufacturer Code:NBP298444AF594
Catalog # NB082715
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant HAAO (NP_064461.1 Met1-Gly286). |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-HAO; 3-HAO 3-hydroxyanthranilate 34-dioxygenase 3-hydroxyanthranilic acid dioxygenase EC 1.13.11.63-hydroxyanthranilate oxygenase HAD HAO; 3-hydroxyanthranilate 3,4-dioxygenase; 3-hydroxyanthranilate oxygenase; 3-hydroxyanthranilic acid dioxygenase; HAAO; HAD
Gene Aliases: 3-HAO; HAAO; HAO
UniProt ID: (Human) P46952
Entrez Gene ID: (Human) 23498
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.