OriGene
Manufacturer Code:CF501257
Catalog # 50-166-0282
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A7 |
Immunogen | Full length human recombit protein of human H6PD produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 6-phosphogluconolactonase; 6PGL; CORTRD1 G6PDH GDH; G6PD, H form; GDH; GDH/6PGL endoplasmic bifunctional protein; glucose 1- dehydrogenase; Glucose 1-dehydrogenase; glucose dehydrogenase; glucose dehyrogenase; Glucose-6-phosphate dehydrogenase; glucose-6-phosphate dehydrogenase, salivary; Hexose-6-phosphate dehydrogenase; hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase)
Gene Aliases: CORTRD1; G6PDH; GDH; H6PD
UniProt ID: (Human) O95479
Entrez Gene ID: (Human) 9563
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.