Invitrogen
Manufacturer Code:PA524960
Catalog # PIPA524960
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 743-769 amino acids from the C-terminal region of human H6PD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6-phosphogluconolactonase; 6PGL; G6PD, H form; G6PDH GDH 6-phosphogluconolactonase G6PD H form GDH/6PGL endoplasmic bifunctional protein glucose 1- dehydrogenase glucose dehydrogenase glucose dehyrogenase glucose-6-phosphate dehydrogenase salivary; GDH; GDH/6PGL endoplasmic bifunctional protein; glucose 1- dehydrogenase; Glucose 1-dehydrogenase; glucose dehydrogenase; glucose dehyrogenase; Glucose-6-phosphate dehydrogenase; glucose-6-phosphate dehydrogenase 1; glucose-6-phosphate dehydrogenase, salivary; Hexose-6-phosphate dehydrogenase; hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase)
Gene Aliases: AI785303; CORTRD1; G6pd1; G6PDH; GDH; Gpd-1; Gpd1; H6PD
UniProt ID: (Human) O95479, (Mouse) Q8CFX1
Entrez Gene ID: (Human) 9563, (Mouse) 100198
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.