R&D Systems
Manufacturer Code:FAB1155N100UG
Catalog # FB1155N100U
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 205326 |
Immunogen | BaF3 mouse pro-B cell line transfected with mouse H60 and mouse myeloma cell line NS0-derived recombinant mouse H60 Asp30-Gln212 Accession No. Q3TDZ7 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Rat Monoclonal Antibody
Protein Aliases: histocompatibility 60; histocompatibility 60a
Gene Aliases: H60
Entrez Gene ID: (Mouse) 15101
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
major histocompatibility complex antigen
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.