Invitrogen
Manufacturer Code:PA5101009
Catalog # PIPA5101009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Histone H3 (mono+di+tri methyl K14). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dimethyl H3 di-methyl H3 dimethyl histone H3 di-methyl histone H3 H3 H3 histone H3 histone family member A H3 histone family member I H3 histone family member K H3 histone family member L H3 histone family member M H3 histone family 2 H3 histone family 3A H3 histone family 3B H3 histone family 3B (H3.3B) H3 histone family 3C H3 K14 H3.1-221 H3.1-291 H3.1-I H3.2 H3.2-221 H3.2-614 H3.2-615 H3.2-616 H3.3A H3.3B H3.5 H3/A H3/b H3/d H3/f H3/i H3/j H3/k H3/l H3/M H3/n H3/o H3-143 H3-291 H3-3A H3-3B H3-53 H3-614 H3a H3b H3-B H3C1 H3c10 H3c11 H3C12 H3C2 H3C3 H3C4 H3C6 H3C7 H3c8 H3f H3-F H3F1K H3F2 H3F3 H3F3A H3F3B H3F3C H3FA H3FB H3FC HIST1H3C H3FD H3FF H3FH H3FI H3FJ H3FK H3FL H3FM H3FN H3g H3h H3i H3K14 Hist1 HIST1H3A HIST1H3B Hist1h3c Hist1h3d HIST1H3E Hist1h3f Hist1h3g Hist1h3h HIST1H3I HIST1H3J HIST2H3A Hist2h3b HIST2H3C Hist2h3c1 Hist2h3c2 Hist2h3c2-ps Hist2h3ca1 Hist2h3ca2 HIST2H3D histone 1 H3a histone 1 H3b histone 1 H3f histone 1 H3h histone 2 H3a histone 2 H3c histone 2 H3c2 histone 2 H3ca2 Histone 3 histone cluster 1 H3a histone cluster 1 H3b histone cluster 1 H3f histone cluster 1 H3h histone cluster 2 H3a histone cluster 2 H3c histone cluster 2 H3c2 histone cluster 2 H3c2 pseudogene histone gene complex 1 histone H3 histone H3.1 Histone H3.2 histone H3.3 Histone H3.3C histone H3.5 Histone H3/a Histone H3/b Histone H3/c Histone H3/d Histone H3/f Histone H3/h Histone H3/i Histone H3/j Histone H3/k histone H3/l Histone H3/m histone H3/o Histone H3K14me1me2me3 histone variant H3.5 M32461 methyl Histone 3 methyl Histone H3 Mono-Di-Tri-Methyl-H3 K14 Mono-Di-Tri-Methyl-H3 Lys14 Mono-Di-Tri-Methyl-Histone H3 K14 Mono-Di-Tri-Methyl-Histone H3 Lys14 Mono-methyl H3 Mono-methyl Histone H3 PP781 Trimethyl H3 Tri-methyl H3 Trimethyl Histone H3 Tri-methyl Histone H3; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3B (H3.3B); H3-clustered histone 13; H3-clustered histone 14; H3-clustered histone 15; H3-clustered histone 2; H3-clustered histone 3; H3-clustered histone 4; H3-clustered histone 6; H3-clustered histone 7; histone 1, H3a; histone 1, H3b; histone 1, H3f; histone 1, H3h; histone 2, H3a; histone 2, H3c; histone 2, H3c2; histone 2, H3ca2; histone cluster 2, H3c2, pseudogene; histone gene complex 1; histone H3; Histone H3.1; Histone H3.2; Histone H3.3; Histone H3.3C; Histone H3.5; Histone H3/a; Histone H3/b; Histone H3/c; Histone H3/d; Histone H3/f; Histone H3/h; Histone H3/i; Histone H3/j; Histone H3/k; Histone H3/l; Histone H3/m; Histone H3/o; histone variant H3.5
Gene Aliases: H3; H3-143; H3-291; H3-3A; H3-3B; H3-5; H3-53; H3-614; H3-B; H3-F; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; H3.3A; H3.3B; H3.5; H3/A; H3/i; H3/k; H3/l; H3/M; H3/n; H3/o; H3a; H3b; H3C1; H3C10; H3C11; H3C12; H3C13; H3C14; H3C15; H3C2; H3C3; H3C4; H3C6; H3C7; H3C8; H3f; H3F1K; H3F2; H3F3; H3F3A; H3F3B; H3F3C; H3FA; H3FB; H3FC HIST1H3C; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; H3FM; H3FN; H3g; H3h; H3i; Hist1; HIST1H3A; HIST1H3B; Hist1h3c; HIST1H3D; HIST1H3E; HIST1H3F; HIST1H3G; HIST1H3H; HIST1H3I; HIST1H3J; HIST2H3A; Hist2h3b; HIST2H3C; Hist2h3c1; Hist2h3c2; Hist2h3c2-ps; Hist2h3ca1; Hist2h3ca2; HIST2H3D; M32461; PP781
UniProt ID: (Human) P84243, (Human) Q6NXT2, (Human) P68431, (Human) Q71DI3, (Mouse) P68433, (Mouse) P84228, (Mouse) P17320
Entrez Gene ID: (Human) 3020, (Human) 3021, (Human) 440093, (Human) 8350, (Human) 8358, (Human) 8968, (Human) 8357, (Human) 333932, (Human) 126961, (Mouse) 360198, (Mouse) 319150, (Mouse) 260423, (Mouse) 319152, (Mouse) 97114
Molecular Function: DNA binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.