Invitrogen
Manufacturer Code:PA5114691
Catalog # PIPA5114691
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human H2B around the acetylation site of Lys5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610022J01Rik Acetyl-Histone H2 Acetyl-Histone H2B K5 Acetyl-Histone H2B Lys5 AV127319 bA317E16.3 dJ160A22.3 dJ193B12.2 dJ221C16.3 dJ221C16.6 dJ221C16.8 dJ97D16.4 GL105 H2B H2B 291A H2B 291B H2b 613 H2B GL105 H2B histone family member M H2B histone family member W testis-specific H2B histone family member W testis specific H2B histone family member A H2B histone family member B H2B histone family member C H2B histone family member D H2B histone family member E H2B histone family member F H2B histone family member J H2B histone family member K H2B histone family member L H2B histone family member M H2B histone family member N H2B histone family member Q H2B histone family member R H2B histone family member S H2B histone family member T H2B histone family member U (testis-specific) H2B histone family member W testis-specific H2B K H2B type 12 H2B.1 H2B.1A H2B.1B H2B.2 H2B/a H2B/b H2B/c H2B/d H2B/e H2B/f H2B/g H2B/h H2B/I H2B/j H2B/k H2B/l H2B/n H2B/q H2B/r H2B/S h2B-143 H2b-221 H2b-291b H2b-613 H2Bb H2BC10 H2BC4 H2BC6 H2BC7 H2BC8 H2b-f H2BFA H2BFAiii H2BFB H2BFC H2BFD H2BFE H2BFF H2BFG H2BFH H2BFJ H2BFK H2BFL H2BFM H2BFN H2BFQ H2BFR H2bfs H2BFT H2BFU H2BFWT H2BGL105 H2BJ H2b-j H2BK H2BK12ac H2BK15ac H2BK5ac H2b-l H2BLys5ac H2b-n H2BQ H2BS14 H2BS14p HIRA-interacting protein 1 HIRA-interacting protein 2 HIRIP1 HIRIP2 Hist1h2ba Hist1h2bb Hist1h2bc Hist1h2bd HIST1H2BE HIST1H2BF HIST1H2BG Hist1h2bh HIST1H2BI HIST1H2BJ Hist1h2bk HIST1H2BL Hist1h2bm HIST1H2BN HIST1H2BO HIST1H3I HIST2H2BE HIST2H2BF Hist3h2bb histone 1 H2ba histone 1 H2bb histone 1 H2bc histone 1 H2bd histone 1 H2bg histone 1 H2bh histone 1 H2bi histone 1 H2bj histone 1 H2bk histone 1 H2bl histone 1 H2bm histone 1 H2bn histone 1 H2bo histone 2 H2be Histone 2B histone 3 H2bb histone cluster 1 H2ba histone cluster 1 H2bb histone cluster 1 H2bc histone cluster 1 H2bd histone cluster 1 H2bg histone cluster 1 H2bh histone cluster 1 H2bi histone cluster 1 H2bj histone cluster 1 H2bk histone cluster 1 H2bl histone cluster 1 H2bm histone cluster 1 H2bn histone cluster 1 H2bo histone cluster 2 H2be histone cluster 2 H2bf histone cluster 3 H2bb histone family member histone H2B histone H2B type 1-A Histone H2B type 1-B histone H2B type 1-C/E/F/G/I Histone H2B type 1-C/E/G histone H2B type 1-D Histone H2B type 1-F/J/L histone H2B type 1-H Histone H2B type 1-J histone H2B type 1-K Histone H2B type 1-L histone H2B type 1-M Histone H2B type 1-N histone H2B type 1-O Histone H2B type 2-E histone H2B type 2-F Histone H2B type 3-B Histone H2B type F-M Histone H2B type F-S histone H2B type F-S-like histone H2B type W-T Histone H2B testis Histone H2B.1 Histone H2B.1 A Histone H2B.1 B histone H2B.2 Histone H2B.a histone H2B.b Histone H2B.c Histone H2B.d histone H2B.e Histone H2B.f Histone H2B.g Histone H2B.h Histone H2B.I Histone H2B.j histone H2B.k Histone H2B.l histone H2B.n Histone H2B.q histone H2B.r Histone H2B.s histone H2B-GL105 LOC102724334 MGC125414 MGC125415 MGC125416 MGC9388 R74621 RP1-193B12.10 STBP T25626 testis-specific histone 2b testis-specific histone H2B Th2b TSH2B TSH2B.1; H2B 291A; H2B 291B; H2b 613; H2B histone family member W testis-specific; H2B histone family, member A; H2B histone family, member B; H2B histone family, member C; H2B histone family, member D; H2B histone family, member E; H2B histone family, member F; H2B histone family, member J; H2B histone family, member K; H2B histone family, member L; H2B histone family, member M; H2B histone family, member N; H2B histone family, member Q; H2B histone family, member R; H2B histone family, member S; H2B histone family, member T; H2B histone family, member U, (testis-specific); H2B histone family, member W, testis-specific; H2B K; H2B type 12; h2B-143; h2B-221; H2B-clustered histone 17; H2B-clustered histone 18; H2B-clustered histone 21; H2B-clustered histone 3; H2B-clustered histone 5; H2B-clustered histone 9; H2B.S histone 1; H2B.U histone 1; H2B.W histone 1; H2B/a; H2B/b; H2B/c; H2B/d; H2B/e; H2B/f; H2B/g; H2B/h; H2B/j; H2B/k; H2B/l; H2B/n; H2B/q; H2B/r; H2B/s; HIRA-interacting protein 1; HIRA-interacting protein 2; histone 1, H2ba; histone 1, H2bb; histone 1, H2bc; histone 1, H2bd; histone 1, H2bg; histone 1, H2bh; histone 1, H2bi; histone 1, H2bj; histone 1, H2bk; histone 1, H2bl; histone 1, H2bm; histone 1, H2bn; histone 1, H2bo; histone 2, H2be; histone 3, H2bb; histone cluster 1, H2bc; histone cluster 1, H2bd; histone cluster 1, H2bh; histone cluster 1, H2bm; histone cluster 1, H2bn; histone cluster 1, H2bo; histone cluster 3, H2bb; histone family member; histone H2B; Histone H2B type 1-A; Histone H2B type 1-B; Histone H2B type 1-C/E/F/G/I; Histone H2B type 1-C/E/G; Histone H2B type 1-D; Histone H2B type 1-F/J/L; Histone H2B type 1-H; Histone H2B type 1-J; Histone H2B type 1-K; Histone H2B type 1-L; Histone H2B type 1-M; Histone H2B type 1-N; Histone H2B type 1-O; Histone H2B type 2-E; Histone H2B type 2-F; Histone H2B type 3-B; Histone H2B type F-M; Histone H2B type F-S; histone H2B type F-S-like; Histone H2B type W-T; Histone H2B, testis; Histone H2B-GL105; Histone H2B.1; Histone H2B.1 A; Histone H2B.1 B; Histone H2B.2; Histone H2B.a; Histone H2B.b; Histone H2B.c; Histone H2B.d; Histone H2B.e; Histone H2B.f; Histone H2B.g; Histone H2B.h; Histone H2B.j; Histone H2B.k; Histone H2B.l; Histone H2B.n; Histone H2B.q; Histone H2B.r; Histone H2B.s; testis-specific histone 2b; Testis-specific histone H2B; TSH2B.1
Gene Aliases: 2610022J01Rik; AV127319; bA317E16.3; dJ160A22.3; dJ193B12.2; dJ221C16.3; dJ221C16.6; dJ221C16.8; dJ97D16.4; GL105; H2B; H2b-143; H2b-221; H2b-291b; H2b-613; H2b-f; H2b-j; H2b-l; H2b-n; H2B.1; H2B.1A; H2B.1B; H2B.2; H2B/a; H2B/b; H2B/c; H2B/d; H2B/e; H2B/f; H2B/j; H2B/k; H2B/l; H2B/n; H2B/r; H2B/S; H2Bb; H2BC1; H2BC10; H2BC11; H2BC12; H2BC13; H2BC14; H2BC15; H2BC17; H2BC18; H2BC21; H2BC3; H2BC4; H2BC5; H2BC6; H2BC7; H2BC8; H2BC9; H2BFA; H2BFAiii; H2BFB; H2BFC; H2BFD; H2BFE; H2BFF; H2BFG; H2BFH; H2BFJ; H2BFK; H2BFL; H2BFM; H2BFN; H2BFQ; H2BFR; H2BFS; H2BFT; H2BFU; H2BFWT; H2BGL105; H2BJ; H2BK; H2BQ; H2BS1; H2BU1; H2BW1; H2BW2; HIRIP1; HIRIP2; HIST1H2BA; HIST1H2BB; HIST1H2BC; HIST1H2BD; HIST1H2BE; HIST1H2BF; HIST1H2BG; HIST1H2BH; HIST1H2BI; HIST1H2BJ; HIST1H2BK; HIST1H2BL; HIST1H2BM; HIST1H2BN; HIST1H2BO; HIST2H2BE; HIST2H2BF; HIST3H2BB; R74621; STBP; T25626; TH2B; TSH2B; TSH2B.1
UniProt ID: (Human) P0C1H6, (Human) Q7Z2G1, (Human) Q96A08, (Human) P33778, (Human) P62807, (Human) Q93079, (Human) P06899, (Human) O60814, (Human) Q99880, (Human) Q99879, (Human) Q99877, (Human) P23527, (Human) Q16778, (Human) Q5QNW6, (Human) Q8N257, (Rat) Q00729, (Mouse) P70696, (Mouse) Q64475, (Mouse) Q6ZWY9, (Mouse) Q64478, (Mouse) P10853, (Mouse) Q8CGP1, (Mouse) P10854, (Mouse) Q64524, (Human) P57053
Entrez Gene ID: (Human) 286436, (Human) 158983, (Human) 255626, (Human) 3018, (Human) 8347, (Human) 3017, (Human) 8339, (Human) 8345, (Human) 8346, (Human) 8970, (Human) 85236, (Human) 8340, (Human) 8342, (Human) 8341, (Human) 8348, (Human) 8349, (Human) 440689, (Human) 128312, (Rat) 24829, (Mouse) 319177, (Mouse) 319178, (Mouse) 68024, (Rat) 361247, (Mouse) 319181, (Rat) 306945, (Mouse) 319182, (Mouse) 319183, (Mouse) 319184, (Rat) 680312, (Mouse) 319186, (Rat) 291157, (Mouse) 319190, (Rat) 691488, (Human) 102724334
Molecular Function: DNA binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.