OriGene
Manufacturer Code:CF803386
Catalog # 50-166-6464
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C2 |
Immunogen | Human recombit protein fragment corresponding to amino acids 2-261 of human H2AFY2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Core histone macro-H2A.2; core histone macroH2A2.2; H2A histone family, member Y2; Histone macroH2A2; macroH2A2; mH2A2
Gene Aliases: H2AFY2; MACROH2A2
UniProt ID: (Human) Q9P0M6
Entrez Gene ID: (Human) 55506
Molecular Function:
DNA binding protein
histone
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.