Proteintech
Manufacturer Code:170301AP150UL
Catalog # 50-173-1337
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | H2AFY2 Fusion Protein Ag10487 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Core histone macro H2A.2 H2A histone family member Y2 H2AFY2 Histone macroH2A2 macroH2A2 mH2A2; Core histone macro-H2A.2; core histone macroH2A2.2; H2A histone family, member Y2; Histone macroH2A2; mH2A2
Gene Aliases: H2AFY2; H2afy3; MACROH2A2; RGD1561371
UniProt ID: (Human) Q9P0M6, (Mouse) Q8CCK0
Entrez Gene ID: (Human) 55506, (Rat) 361844, (Mouse) 404634
Molecular Function: DNA binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.