Invitrogen
Manufacturer Code:PA5109802
Catalog # PIPA5109802
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-128 of human H2AFV (NP_036544.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C530002L11Rik H2A.F/Z H2A.Z-2 H2afv H2av Histone H2A.V; H2A histone family, member V; H2A.F/Z; H2A.Z variant histone 2; histone H2A.F/Z; Histone H2A.V; purine-rich binding element protein B
Gene Aliases: C530002L11Rik; H2A.Z-2; H2AFV; H2AV; H2AZ2
UniProt ID: (Human) Q71UI9, (Mouse) Q3THW5
Entrez Gene ID: (Human) 94239, (Mouse) 77605, (Rat) 685909
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.