Invitrogen
Manufacturer Code:PA5104550
Catalog # PIPA5104550
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Histone H1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C86609 H1 histone family member O oocyte specific H1 histone family member O oocyte-specific H1.8 H1fo H1foo H1OO Histone H1oo oocyte-specific histone H1 Oocyte-specific linker histone H1 osH1 RGD1566236; H1 histone family, member O, oocyte-specific; Histone H1.8; Histone H1oo; Oocyte-specific histone H1; Oocyte-specific linker histone H1; osH1
Gene Aliases: H1-8; H1.8; H1FOO; H1OO; OSH1; RGD1566236
UniProt ID: (Human) Q8IZA3
Entrez Gene ID: (Human) 132243, (Rat) 502875
Molecular Function: DNA binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.