Invitrogen
Manufacturer Code:PA581349
Catalog # PIPA581349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Mouse CDH13/Cadherin-13/H Cadherin protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cadherin-13; Cadherin-13 Truncated cadherin T-cadherin T-cad Heart cadherin H-cadherin P105 CDH13 CDHH; H-cadherin; Heart cadherin; T-cad; Truncated cadherin
Gene Aliases: 4932416G01Rik; Cdh13; Cdht; T-cadherin; Tcad
UniProt ID: (Mouse) Q9WTR5
Entrez Gene ID: (Mouse) 12554
Molecular Function: cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.