Invitrogen
Manufacturer Code:PA5114998
Catalog # PIPA5114998
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CDH13 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4932416G01Rik CAD13 cadherin 13 cadherin 13 H-cadherin (heart) cadherin-13 CD39 antigen-like 3 CD39L3 CD39-like 3 CDH13 CDHH Cdht ecto-apyrase 3 ecto-ATPase 3 ecto-ATPDase 3 HB6 H-cadherin H-cadherin (heart) heart cadherin heart-cadherin NTPDase 3 NTPDase-3 P105 T cad T Cadherin Tcad T-cad T-cadherin Truncated cadherin truncated-cadherin; cadherin 13, H-cadherin (heart); Cadherin-13; H-cadherin; H-cadherin (heart); Heart cadherin; P105; T-cad; T-cadherin; Truncated cadherin
Gene Aliases: 4932416G01Rik; CDH13; CDHH; Cdht; P105; T-cadherin; Tcad
UniProt ID: (Human) P55290, (Mouse) Q9WTR5
Entrez Gene ID: (Human) 1012, (Mouse) 12554
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.