Invitrogen
Manufacturer Code:PA579395
Catalog # PIPA579395
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human GTPase HRAS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-H-ras; c-Ha-ras p21 protein; c-Ha-ras transgene; c-has/bas p21 protein; c-ras-Ki-2 activated oncogene; GTP- and GDP-binding peptide B; GTPase HRas; GTPase HRas H-Ras-1 Ha-Ras Transforming protein p21 c-H-ras p21ras GTPase HRas N-terminally processed HRAS HRAS1; GTPase HRas, N-terminally processed; H-ras 1 protein; H-Ras-1; Ha-Ras; Ha-Ras1 proto-oncoprotein; Harvey ras1 protein; Harvey rat sarcoma viral (v-Ha-ras) oncogene homolog; Harvey rat sarcoma viral oncogene homolog; Harvey rat sarcoma viral oncoprotein; Harvey rat sarcoma virus oncogene 1; p19 H-RasIDX protein; p21ras; Ras family small GTP binding protein H-Ras; transformation gene: oncogene HAMSV; Transforming protein p21; v-Ha-ras Harvey rat sarcoma viral oncogene homolog
Gene Aliases: C-BAS/HAS; C-H-RAS; c-Ha-ras; C-HA-RAS1; c-rasHa; CTLO; H-ras; H-RASIDX; Ha-ras; HAMSV; Harvey-ras; HRAS; Hras-1; HRAS1; Kras2; p21ras; ras; RASH1
UniProt ID: (Human) P01112, (Mouse) Q61411, (Rat) P20171
Entrez Gene ID: (Human) 3265, (Mouse) 15461, (Rat) 293621
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.