Proteintech
Manufacturer Code:270791AP150UL
Catalog # 50-173-5553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Growth hormone Fusion Protein Ag22841 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GH; GH GH N GH1 GHN Growth hormone growth hormone 1 hGH N Pituitary growth hormone Somatotropin; Growth hormone; Growth hormone 1; growth hormone B5; Pituitary growth hormone; Somatotropin
Gene Aliases: GH; GH-N; GH1; GHB5; GHN; hGH-N; IGHD1B
UniProt ID: (Human) P01241
Entrez Gene ID: (Human) 2688
Molecular Function:
growth factor
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.