Invitrogen
Manufacturer Code:PA579309
Catalog # PIPA579309
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of mouse Growth hormone receptoR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GH receptor; GH-binding protein; Growth hormone receptor; Growth hormone receptor GH receptor Somatotropin receptor Growth hormone-binding protein GH-binding protein GHBP Serum-binding protein Ghr; Growth hormone receptor precursor (GH receptor) (GH binding protein) (GHBP) (Serum binding protein); growth hormone receptor precursor splice variant D56; growth hormone receptor variant d5-6; growth hormone receptor/binding protein; Growth hormone-binding protein; Serum-binding protein; Somatotropin receptor
Gene Aliases: GHBP; Ghr; GHR/BP
UniProt ID: (Rat) P16310, (Mouse) P16882
Entrez Gene ID: (Rat) 25235, (Mouse) 14600
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.