R&D Systems
Manufacturer Code:AF1360SP
Catalog # AF1360SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Growth Hormone R/GHR Thr25-Gln273 Accession # Q3UP14 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: GH receptor; GH receptor GHBP GHR growth hormone binding protein growth hormone receptor serum binding protein Somatotropin receptor; GH-binding protein; growth hormone binding protein; Growth hormone receptor; Growth hormone-binding protein; serum binding protein; Serum-binding protein; Somatotropin receptor
Gene Aliases: GHBP; GHIP; GHR
UniProt ID: (Human) P10912
Entrez Gene ID: (Human) 2690
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.