Novus Biologicals
Manufacturer Code:NBP266893
Catalog # NBP266893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JM108-09 |
Immunogen | Full length protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: GH; GHIGHD1B GHNGrowth hormone GH-Nsomatotropin growth hormone 1Pituitary growth hormone hGH-N; Growth hormone; Growth hormone 1; growth hormone B5; Pituitary growth hormone; Somatotropin
Gene Aliases: GH; GH-N; GH1; GHB5; GHN; hGH-N; IGHD1B
UniProt ID: (Human) P01241
Entrez Gene ID: (Human) 2688
Molecular Function:
growth factor
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.