Invitrogen
Manufacturer Code:MA511926
Catalog # PIMA511926
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Type | Antibody |
Clone | GH01 (No.2),GH02 (No.7) |
Immunogen | Purified recombit human growth hormone |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Antibody Cocktail
For Research Use Only
Protein Aliases: GH; Growth hormone; Growth hormone 1; growth hormone B5; Pituitary growth hormone; pituitary growth hormone somatotropin; Somatotropin
Gene Aliases: GH; GH-N; GH1; GHB5; GHN; hGH-N; IGHD1B
UniProt ID: (Human) P01241
Entrez Gene ID: (Human) 2688
Molecular Function: growth factor peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.