R&D Systems
Manufacturer Code:MAB1067500
Catalog # MAB1067500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 178902 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Growth Hormone Phe27-Phe217 Accession # CAA23779 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GH; GH1 GHB5 GHIGHD1B GHN GH-N GHNGrowth hormone GH-Nsomatotropin growth hormone 1Pituitary growth hormone hGH-N IGHD1A IGHD2 Somatotropin; Growth hormone; Growth hormone 1; growth hormone B5; Pituitary growth hormone; Somatotropin
Gene Aliases: GH; GH-N; GH1; GHB5; GHN; hGH-N; IGHD1B
UniProt ID: (Human) P01241
Entrez Gene ID: (Human) 2688
Molecular Function:
growth factor
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.