Abnova
Manufacturer Code:PAB9705
Catalog # 89-134-347
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to N-terminus rat Grin2c. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C on dry atmosphere. After reconstitution with 50&muL of PBS,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Grin2c.
For Research Use Only
Protein Aliases: GluN2C; Glutamate [NMDA] receptor subunit epsilon-3; Glutamate receptor ionotropic, NMDA 2C; glutamate receptor, ionotropic, NMDA2C; N-methyl D-aspartate receptor subtype 2C; NMDA glutamate receptor; NMDAR2C; NR2C
Gene Aliases: GluN2C; Grin2c; NR2C
UniProt ID: (Rat) Q00961
Entrez Gene ID: (Rat) 24411
Molecular Function: ion channel ionotropic glutamate receptor ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.