Abnova
Manufacturer Code:PAB9641
Catalog # 89-134-295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding S1480 of rat Grin2b. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Grin2b.
For Research Use Only
Protein Aliases: GluN2B; Glutamate [NMDA] receptor subunit epsilon-2; Glutamate receptor ionotropic, NMDA 2B; glutamate receptor, ionotropic, NMDA2B; N-methyl D-aspartate receptor subtype 2B; NMDAR2B; NR2B
Gene Aliases: GluN2B; Grin2b
UniProt ID: (Rat) Q00960
Entrez Gene ID: (Rat) 24410
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.