Abnova
Manufacturer Code:MAB11715
Catalog # 89-109-849
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S308-48 |
Immunogen | Recombinant fusion protein corresponding to amino acids 42-361 at extracellular N-terminus of rat Grin1. |
Conjugate | ATTO 633 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial synthetic protein of Grin1.
For Research Use Only
Protein Aliases: GluN1; Glutamate [NMDA] receptor subunit zeta-1; Glutamate receptor ionotropic, NMDA 1; N-methyl-D-aspartate glutamate receptor; N-methyl-D-aspartate receptor subunit NR1; neurotransmitter receptor; NMD-R1; NMDA R1 receptor C1 cassette; NMDAR1; NR1
Gene Aliases: GluN1; Grin1; NMDAR1; NR1
UniProt ID: (Rat) P35439
Entrez Gene ID: (Rat) 24408
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.