Invitrogen
Manufacturer Code:PA520452
Catalog # PIPA520452
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 19 amino acid peptide near the carboxy terminus of the human Grik4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EAA1; EAA1 GRIK KA1 excitatory amino acid receptor 1 glutamate receptor KA-1 glutamate receptor KA1 glutamate receptor ionotropic kainate 4; Excitatory amino acid receptor 1; GluK4; Glutamate receptor ionotropic, kainate 4; Glutamate receptor KA-1; glutamate receptor KA1; glutamate receptor, ionotropic, kainate 4; KA1; kainate receceptor
Gene Aliases: 6330551K01Rik; EAA1; GluK4; GluRgamma1; GRIK; GRIK4; KA-1; KA1
UniProt ID: (Human) Q16099, (Rat) Q01812, (Mouse) Q8BMF5
Entrez Gene ID: (Human) 2900, (Rat) 24406, (Mouse) 110637
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.