Abnova
Manufacturer Code:MAB11655
Catalog # 89-109-807
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S21-32 |
Immunogen | Recombinant fusion protein corresponding to amino acids 834-883 of rat Gria2. |
Conjugate | Streptavidin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial synthetic protein of Gria2.
For Research Use Only
Protein Aliases: AMPA-selective glutamate receptor 2; GluA2; GluR-2; GluR-B; GluR-K2; GluR2; Glutamate receptor 2; glutamate receptor B; Glutamate receptor ionotropic, AMPA 2
Gene Aliases: GluA2; gluR-B; GluR-K2; GluR2; Gria2
UniProt ID: (Rat) P19491
Entrez Gene ID: (Rat) 29627
Molecular Function: ion channel ionotropic glutamate receptor ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.