R&D Systems
Manufacturer Code:MAB51901100
Catalog # MAB51901100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 955323 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Gremlin Met1-Asp184 Accession &num O60565 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cell proliferation-inducing gene 2 protein; Cell proliferation-inducing gene 2 protein CKTSF1B1gremlin 1 cysteine knot superfamily homolog Cysteine knot superfamily 1 BMP antagonist 1gremlin 1 cysteine knot superfamily homolog (Xenopus laevis) DAN domain family member 2 DAND2 DAND2GREMLIN Down-regulated in Mos-transformed cells protein DRM DRMMGC126660 GREM1 gremlin gremlin 1 gremlin 1-like protein gremlin-1 IHG-2 Increased in high glucose protein 2 increased in high glucose-2 proliferation-inducing gene 2; colorectal adenoma and carcinoma 1; Cysteine knot superfamily 1, BMP antagonist 1; DAN domain family member 2; Down-regulated in Mos-transformed cells protein; gremlin 1, cysteine knot superfamily, homolog; gremlin 1-like protein; Gremlin-1; IHG-2; Increased in high glucose protein 2; increased in high glucose-2
Gene Aliases: C15DUPq; CKTSF1B1; CRAC1; CRCS4; DAND2; DRM; DUP15q; GREM1; GREMLIN; HMPS; HMPS1; IHG-2; MPSH; PIG2
UniProt ID: (Human) O60565
Entrez Gene ID: (Human) 26585
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.