Novus Biologicals
Manufacturer Code:NB10056883
Catalog # NB10056883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide containing amino acids QPGSRNRGRGQGRGTA (52-67) of human CKTSF1B1 (NM_013372). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell proliferation-inducing gene 2 protein; Cell proliferation-inducing gene 2 protein CKTSF1B1gremlin 1 cysteine knot superfamily homolog Cysteine knot superfamily 1 BMP antagonist 1gremlin 1 cysteine knot superfamily homolog (Xenopus laevis) DAN domain family member 2 DAND2GREMLIN Down-regulated in Mos-transformed cells protein DRMMGC126660 gremlin gremlin 1 gremlin 1-like protein gremlin-1 IHG-2 Increased in high glucose protein 2 increased in high glucose-2 proliferation-inducing gene 2; colorectal adenoma and carcinoma 1; Cysteine knot superfamily 1, BMP antagonist 1; DAN domain family member 2; Down-regulated in Mos-transformed cells protein; gremlin 1, cysteine knot superfamily, homolog; gremlin 1-like protein; Gremlin-1; IHG-2; Increased in high glucose protein 2; increased in high glucose-2
Gene Aliases: C15DUPq; CKTSF1B1; CRAC1; CRCS4; DAND2; DRM; DUP15q; GREM1; GREMLIN; HMPS; HMPS1; IHG-2; MPSH; PIG2
UniProt ID: (Human) O60565
Entrez Gene ID: (Human) 26585
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.