Invitrogen
Manufacturer Code:48583182
Catalog # 50-112-4178
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GzA-3G8.5 |
Conjugate | eFluor 450 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Autocrine thymic lymphoma granzyme-like serine protease; BLT esterase; CTLA-3; CTLA3 HFSP; Fragmentin-1; Granzyme A; H factor; Hanukah factor; serine esterase 1; T cell-specific serine protease 1; TSP-1
Gene Aliases: AW494114; Ctla-3; Ctla3; Gzma; Hf; Mtsp-1; SE1; TSP-1; TSP1
UniProt ID: (Mouse) P11032
Entrez Gene ID: (Mouse) 14938
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.