R&D Systems
Manufacturer Code:MAB29051
Catalog # MAB29051
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 356412 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Granzyme A Cys26-Val262 Accession # P12544 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CTL tryptase; CTL tryptase CTLA3 CTLA3Granzyme A (Cytotoxic T-lymphocyte-associated serine esterase-3 Hanukah factorserine protease) Cytotoxic T-lymphocyte proteinase 1 EC 3.4.21 fragmentin-1 granzyme A granzyme A (granzyme 1 cytotoxic T-lymphocyte-associated serine esterase 3) Granzyme-1 GZMA H factor Hanukkah factor HF HFSP HFSPEC 3.4.21.78; Cytotoxic T-lymphocyte proteinase 1; Cytotoxic T-lymphocyte-associated serine esterase-3; Fragmentin-1; granzyme 1; Granzyme A; Granzyme A (Cytotoxic T-lymphocyte-associated serine esterase-3; Hanukah factor serine protease); granzyme A (granzyme 1, cytotoxic T-lymphocyte-associated serine esterase 3); Granzyme-1; H factor; Hanukah factor serine protease); Hanukkah factor; HF
Gene Aliases: CTLA3; GZMA; HFSP
UniProt ID: (Human) P12544
Entrez Gene ID: (Human) 3001
Molecular Function: annexin calcium-binding protein calmodulin enzyme modulator hydrolase intracellular calcium-sensing protein peptide hormone protease protease inhibitor receptor serine protease signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.