Invitrogen
Manufacturer Code:710191
Catalog # PI710191
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Clone | 2HCLC |
Immunogen | Peptide corresponding to amino acids 573588 of human PC cell-derived growth factor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: Acrogranin; Epithelin precursor; Epithelin-1; Epithelin-2; GEP GP88 PCDGF PEPI PGRN PC cell-derived growth factor acrogranin granulin-epithelin granulins proepithelin progranulin; Glycoprotein of 88 Kda; GP88; Granulin A; Granulin B; Granulin C; Granulin D; Granulin E; Granulin F; Granulin G; Granulin precursor; Granulin-1; Granulin-2; Granulin-3; Granulin-4; Granulin-5; Granulin-6; Granulin-7; granulin-epithelin; Paragranulin; PC cell-derived growth factor; PCDGF; PEPI; PGRN; Proepithelin; Progranulin
Gene Aliases: CLN11; GEP; GP88; GRN; PCDGF; PEPI; PGRN
UniProt ID: (Human) P28799
Entrez Gene ID: (Human) 2896
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.