Invitrogen
Manufacturer Code:OSR00147W
Catalog # PIOSR00147W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from c-terminal region of mouse ocular albinism type 1 protein homolog |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G-protein coupled receptor 143; G-protein coupled receptor 143 ocular albinism type 1 protein homolog MOA1 Gpr143 Oa1; homolog of human ocular albinism 1 (Nettleship-Falls); MOA1; Ocular albinism type 1 protein homolog
Gene Aliases: Gpr143; Oa1
UniProt ID: (Mouse) P70259
Entrez Gene ID: (Mouse) 18241
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.