Abnova
Manufacturer Code:PAB12139
Catalog # 89-115-637
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 100-170 of mouse Gpihbp1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Gpihbp1.
For Research Use Only
Protein Aliases: 1110002J19Rik; Glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein 1; GPI-HBP1; gpihbp1; High density lipoprotein-binding protein 1
Gene Aliases: 1110002J19Rik; GPI-HBP1; Gpihbp1; Hbp1
UniProt ID: (Mouse) Q9D1N2
Entrez Gene ID: (Mouse) 68453
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.