Invitrogen
Manufacturer Code:PA116976
Catalog # 01-670-153
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 100-170 of mouse GPI-anchored HDL-binding protein 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein 1; GPI-anchored HDL-binding protein 1; GPI-anchored HDL-binding protein 1 antibody GPI-anchored High-Density Lipid-binding protein 1 antibody GlycosylPhosphatidylInositol-anchored high-de; GPI-HBP1; gpihbp1; High density lipoprotein-binding protein 1
Gene Aliases: 1110002J19Rik; GPI-HBP1; Gpihbp1; Hbp1; RGD1564237
UniProt ID: (Mouse) Q9D1N2
Entrez Gene ID: (Rat) 300027, (Mouse) 68453
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.