Invitrogen
Manufacturer Code:PA19000
Catalog # 01-674-584
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Internal sequence corresponding to amino acids CLREQGRGKDQPGRL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Formimidoyltetrahydrofolate cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; Formiminotransferase-cyclodeaminase; FTCD; FTCD LCHC1 formiminotransferase cyclodeaminase formimidoyltransferase cyclodeaminase; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; LCHC1
Gene Aliases: FTCD; LCHC1
UniProt ID: (Human) O95954
Entrez Gene ID: (Human) 10841
Molecular Function:
deaminase
hydrolase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.