Invitrogen
Manufacturer Code:PA128666
Catalog # PIPA128666
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Chinese hamster |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Secondary Antibody |
Immunogen | Full length native hamster whole (purified). |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. Store in the dark. |
Target | IgG |
Goat Polyclonal Secondary Antibody
For Research Use Only
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.