Abnova
Manufacturer Code:MAB1886
Catalog # 89-110-690
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | R4.5 |
Immunogen | Native purified rat Gnai1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against native Gnai1.
For Research Use Only
Protein Aliases: Adenylate cyclase-inhibiting G alpha protein; BPGTPB; guanine nucleotide binding protein (G protein), alpha inhibiting 1; guanine nucleotide binding protein, alpha inhibiting 1; Guanine nucleotide-binding protein G(i) subunit alpha-1; guanine nucleotide-binding protein G(i), alpha-1 subunit
Gene Aliases: BPGTPB; Gnai-1; Gnai1
UniProt ID: (Rat) P10824
Entrez Gene ID: (Rat) 25686
Molecular Function: G-protein enzyme modulator heterotrimeric G-protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.