Novus Biologicals
Manufacturer Code:NBP189749
Catalog # NBP189749
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:QHWSYGLRPGGKRDAENLIDSFQEIVKEVGQLAETQRFECTTHQPRSPLRDLKGALESLIEEETGQKKI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GNRH GnRH-associated peptide 1 gonadotropin-releasing hormone 1 (leutinizing-releasing hormone) gonadotropin-releasing hormone 1 (luteinizing-releasing hormone) GRH LHRH LNRH luliberin I Progonadoliberin I progonadoliberin-1 prolactin release-inhibiting factor; GnRH-associated peptide 1; GnRH-associated peptide I; GnRH-I; Gonadoliberin I; Gonadoliberin-1; Gonadorelin; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); Gonadotropin-releasing hormone I; leuteinizing-releasing hormone; LH-RH I; Luliberin I; Luteinizing hormone-releasing hormone I; Progonadoliberin I; Progonadoliberin-1; prolactin release-inhibiting factor
Gene Aliases: GNRH; GNRH1; GRH; LHRH; LNRH
UniProt ID: (Human) A0AVP0
Entrez Gene ID: (Human) 2796
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.