Invitrogen
Manufacturer Code:MA516368
Catalog # PIMA516368
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SP86 |
Immunogen | Synthetic peptide derived from C-terminus of human glypican-3 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: DGSX GTR2-2 MXR7 OCI-5 SDYS SGB SGBS SGBS1 glypican proteoglycan 3 glypican-3 heparan sulphate proteoglycan intestinal protein OCI-5 secreted glypican-3; glypican proteoglycan 3; Glypican-3; Glypican-3 alpha subunit; Glypican-3 beta subunit; GTR2-2; heparan sulphate proteoglycan; Intestinal protein OCI-5; MXR7; secreted glypican-3
Gene Aliases: DGSX; GPC3; GTR2-2; MXR7; OCI-5; OCI5; SDYS; SGB; SGBS; SGBS1
UniProt ID: (Human) P51654
Entrez Gene ID: (Human) 2719
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.