Invitrogen
Manufacturer Code:MA531552
Catalog # PIMA531552
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT2473 |
Immunogen | Recombit protein encompassing a sequence within the C-terminus region of human Glypican-3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DGSX antibody GPC3 antibody GTR22 antibody Glypican-3 antibody &lsqbGT2473] antibody Glypican-3 antibody MXR7 antibody OCI5 antibody SDYS antibody SGB antibody SGBS antibody SGBS1 antibody glypican 3 antibody; glypican proteoglycan 3; Glypican-3; Glypican-3 alpha subunit; Glypican-3 beta subunit; GTR2-2; heparan sulphate proteoglycan; Intestinal protein OCI-5; MXR7; secreted glypican-3
Gene Aliases: DGSX; GPC3; GTR2-2; MXR7; OCI-5; OCI5; SDYS; SGB; SGBS; SGBS1
UniProt ID: (Human) P51654
Entrez Gene ID: (Human) 2719
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.