Invitrogen
Manufacturer Code:MA530614
Catalog # PIMA530614
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 12 |
Immunogen | Recombit Mouse Glypican 3/GPC3/OCI-5 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Glypican-3; Glypican-3 &lsqbPrecursor] Intestinal protein OCI-5 GTR2-2 MXR7 GPC3 OCI5; Glypican-3 alpha subunit; Glypican-3 beta subunit
Gene Aliases: Gpc3; OCI-5
UniProt ID: (Mouse) Q8CFZ4
Entrez Gene ID: (Mouse) 14734
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.