Invitrogen
Manufacturer Code:PA577986
Catalog # PIPA577986
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the C-terminus region of human GPC3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DGSX GPC3 GPC3 GTR22 MXR7 OCI5 SDYS SGB SGBS SGBS1 glypican 3; glypican proteoglycan 3; Glypican-3; Glypican-3 alpha subunit; Glypican-3 beta subunit; GTR2-2; heparan sulphate proteoglycan; Intestinal protein OCI-5; MXR7; secreted glypican-3
Gene Aliases: DGSX; GPC3; GTR2-2; MXR7; OCI-5; OCI5; SDYS; SGB; SGBS; SGBS1
UniProt ID: (Human) P51654
Entrez Gene ID: (Human) 2719
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.