Proteintech
Manufacturer Code:167001AP150UL
Catalog # 50-173-1124
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Glypican 1 Fusion Protein Ag10095 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glypican Glypican 1 GPC1; glypican proteoglycan 1; Glypican-1; GPI-anchored cell surface heparan sulfate proteoglycan; HSPG M12; Secreted glypican-1
Gene Aliases: AI462976; glypican; GPC1; HSPG M12; HSPGM12
UniProt ID: (Human) P35052, (Mouse) Q9QZF2, (Rat) P35053
Entrez Gene ID: (Human) 2817, (Mouse) 14733, (Rat) 58920
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.