R&D Systems
Manufacturer Code:AF5944SP
Catalog # AF5944SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Glyoxalase II Met49-Ile254 Accession # Q16775 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.2.6 GLO2hydroxyacyl glutathione hydrolase Glx II GLX2 GLXII Glyoxalase II HAGH HAGH1GLX2 hydroxyacylglutathione hydrolase hydroxyacylglutathione hydrolase mitochondrial hydroxyacylglutathione hydroxylase; Glx II; Glyoxalase II; Hydroxyacylglutathione hydrolase, mitochondrial; hydroxyacylglutathione hydroxylase
Gene Aliases: GLO2; GLX2; GLXII; HAGH; HAGH1
UniProt ID: (Human) Q16775
Entrez Gene ID: (Human) 3029
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.