Novus Biologicals
Manufacturer Code:NBP259369
Catalog # NBP259369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 9F11 |
Immunogen | Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Aldoketomutase; Aldoketomutase EC 4.4.1.5 GLOD1 Glx I GLYI glyoxalase domain containing 1 glyoxalase Ialdoketomutase Ketone-aldehyde mutase lactoyl glutathione lyase lactoylglutathione lyase methylglyoxalase S-D-lactoylglutathione methylglyoxal lyase; epididymis secretory protein Li 74; Glx I; glyoxalase domain containing 1; Glyoxalase I; Ketone-aldehyde mutase; lactoyl glutathione lyase; Lactoylglutathione lyase; Methylglyoxalase; S-D-lactoylglutathione methylglyoxal lyase
Gene Aliases: GLO1; GLOD1; GLYI; HEL-S-74
UniProt ID: (Human) Q96J41
Entrez Gene ID: (Human) 2739
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.