Novus Biologicals
Manufacturer Code:NBP290549AF405
Catalog # NB074820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 43 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Mouse Glyoxalase I (NP_079650.3 Ala 2-Ile 184). |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Aldoketomutase; Aldoketomutase EC 4.4.1.5 GLOD1 Glx I GLYI glyoxalase domain containing 1 glyoxalase Ialdoketomutase Ketone-aldehyde mutase lactoyl glutathione lyase lactoylglutathione lyase methylglyoxalase S-D-lactoylglutathione methylglyoxal lyase; epididymis secretory protein Li 74; Glx I; glyoxalase domain containing 1; Glyoxalase I; Ketone-aldehyde mutase; lactoyl glutathione lyase; Lactoylglutathione lyase; Methylglyoxalase; S-D-lactoylglutathione methylglyoxal lyase
Gene Aliases: GLO1; GLOD1; GLYI; HEL-S-74
UniProt ID: (Human) Q96J41
Entrez Gene ID: (Human) 2739
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.