Novus Biologicals
Manufacturer Code:NB600611
Catalog # NB600611
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Human Muscle Glycogen Synthase phospho peptide corresponding to a Ser640 region of the human protein conjugated to Keyhole Limpet Hemocyanin (KLH). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.4.1.11 glycogen [starch] synthase muscle glycogen synthase 1 (muscle) GYSGSY; Glycogen [starch] synthase, muscle; glycogen synthase 1 (muscle)
Gene Aliases: GSY; GYS; GYS1
UniProt ID: (Human) P13807
Entrez Gene ID: (Human) 2997
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.