Novus Biologicals
Manufacturer Code:NB1107378H
Catalog # NB1107378H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8G7 |
Immunogen | Recombinant full length protein (Human). |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Brain glycogen phosphorylase EC 2.4.1.1 Glycogen phosphorylase B Glycogen phosphorylase brain form Glycogen Phosphorylase Isoenzyme BB MGC9213 Phosphorylase glycogen brain PYGB; glycogen phosphorylase B; Glycogen phosphorylase, brain form
Gene Aliases: GPBB; PYGB
UniProt ID: (Human) P11216
Entrez Gene ID: (Human) 5834
Molecular Function: phosphorylase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.