Novus Biologicals
Manufacturer Code:NBP130930
Catalog # NBP130930
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Glycine Receptor alpha 3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glycine receptor alpha 3 subunit glycine receptor subunit alpha-3 glycine receptor alpha 3 glycine receptor alpha-3 polypeptide ligand gated ion channel; Glycine receptor subunit alpha-3; glycine receptor, alpha 3; glycine receptor, alpha-3 polypeptide; ligand gated ion channel
Gene Aliases: GLRA3
UniProt ID: (Human) O75311
Entrez Gene ID: (Human) 8001
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.