Novus Biologicals
Manufacturer Code:NBP271838PECY55
Catalog # N271838PEC5
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8A3 |
Immunogen | Full length recombinant protein of human Glycine N-Methyltransferase/GNMT (NP_061833) produced in HEK293T cells. |
Conjugate | PE-Cyanine5.5 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: EC 2.1.1.20 glycine N-methyltransferase; epididymis secretory sperm binding protein Li 182mP; Glycine N-methyltransferase
Gene Aliases: GNMT; HEL-S-182mP
UniProt ID: (Human) Q14749
Entrez Gene ID: (Human) 27232
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.