R&D Systems
Manufacturer Code:MAB6526
Catalog # MAB6526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 691305 |
Immunogen | <em>E. coli</em>-derived recombinant human Glycine N-methyltransferase/GNMT Met1-Asp295 Accession # Q14749 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.1.1.20 glycine N-methyltransferase Glycine NMethyltransferase GNMT; epididymis secretory sperm binding protein Li 182mP; Glycine N-methyltransferase
Gene Aliases: GNMT; HEL-S-182mP
UniProt ID: (Human) Q14749
Entrez Gene ID: (Human) 27232
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.